Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCK Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $456.00
Specifications
Antigen | Cholecystokinin |
---|---|
Clone | 2753A |
Dilution | Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 3-25 ug/mL |
Applications | Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11040SP
![]() |
R&D Systems
MAB11040SP |
25 μg |
Each for $200.50
|
|
|||||
MAB11040100
![]() |
R&D Systems
MAB11040100 |
100 μg |
Each for $456.00
|
|
|||||
Description
Cholecystokinin Monoclonal antibody specifically detects Cholecystokinin in Human samples. It is validated for Immunohistochemistry, ImmunocytochemistrySpecifications
Cholecystokinin | |
Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 3-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
cholecystokinin, MGC117187 | |
Human CCK synthetic peptide, Accession # P06307 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2753A | |
Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
P06307 | |
885 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title