Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL2/JE/MCP-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 14 publications
$214.00 - $2020.00
Specifications
Antigen | CCL2/JE/MCP-1 |
---|---|
Clone | 24822 |
Dilution | Western Blot 1 ug/mL, Neutralization 0.9-4.5 ug/mL |
Applications | Western Blot, Neutralization |
Classification | Monoclonal |
Description
CCL2/JE/MCP-1 Monoclonal specifically detects CCL2/JE/MCP-1 in Human samples. It is validated for Western Blot, Neutralization.Specifications
CCL2/JE/MCP-1 | |
Western Blot 1 ug/mL, Neutralization 0.9-4.5 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, MCP-1, SCYA2, SMC-CF | |
CCL2 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
24822 | |
Western Blot, Neutralization | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
6347 | |
E. coli-derived recombinant human CCL2/JE/MCP-1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CCL2/JE/MCP-1 in Western blots. Does not cross-react with recombinant human (rh) CCL1, 3, 4, 5, 7, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, rmCCL1, 2, 3, 4, 5, 6, 7, 9/10MIP-1 gamma , 11, 12, 17, 19, 20, 22, 24, 25, or rrCCL20. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title