Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL27/CTACK Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Supplier: R&D Systems MAB3761
This item is not returnable.
View return policy
Description
CCL27/CTACK Monoclonal specifically detects CCL27/CTACK in Human samples. It is validated for Immunohistochemistry, ELISA Capture (Matched Antibody Pair).Specifications
CCL27/CTACK | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ALP, C-C motif chemokine 27, chemokine (C-C motif) ligand 27, CTACK, CTACKSmall-inducible cytokine A27, CTAK, cutaneous T-cell attracting chemokine, Cutaneous T-cell-attracting chemokine, ESKINE, IL-11 Ralpha-locus chemokine, ILC, ILCCC chemokine ILC, PESKY, SCYA27IL-11 R-alpha-locus chemokine, Skinkine, small inducible cytokine subfamily A (Cys-Cys), member 27 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10850 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Immunohistochemistry, ELISA | |
124302 | |
Immunohistochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
NP_006655 | |
CCL27 | |
E. coli-derived recombinant human CCL27/CTACK Phe25-Gly112 Accession # NP_006655 | |
500 μg | |
Primary | |
Detects human CCL27/CTACK in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse CCL27, recombinant human (rh) CCL25, or rhCCL19 is observed. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction