Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL27/CTACK Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
$200.50 - $667.00
Specifications
Antigen | CCL27/CTACK |
---|---|
Clone | 124302 |
Dilution | Immunohistochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Immunohistochemistry, ELISA |
Classification | Monoclonal |
Description
CCL27/CTACK Monoclonal specifically detects CCL27/CTACK in Human samples. It is validated for Immunohistochemistry, ELISA Capture (Matched Antibody Pair).Specifications
CCL27/CTACK | |
Immunohistochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ALP, C-C motif chemokine 27, chemokine (C-C motif) ligand 27, CTACK, CTACKSmall-inducible cytokine A27, CTAK, cutaneous T-cell attracting chemokine, Cutaneous T-cell-attracting chemokine, ESKINE, IL-11 Ralpha-locus chemokine, ILC, ILCCC chemokine ILC, PESKY, SCYA27IL-11 R-alpha-locus chemokine, Skinkine, small inducible cytokine subfamily A (Cys-Cys), member 27 | |
CCL27 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
124302 | |
Immunohistochemistry, ELISA | |
Unconjugated | |
Mouse | |
Human | |
NP_006655 | |
10850 | |
E. coli-derived recombinant human CCL27/CTACK Phe25-Gly112 Accession # NP_006655 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CCL27/CTACK in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse CCL27, recombinant human (rh) CCL25, or rhCCL19 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title