Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human CCL3/MIP-1 alpha DuoSet ELISA

Catalog No. p-4754101
Click to view available options
Format:
15 96-well Plates
5 96-well Plates
This item is not returnable. View return policy
This item is not returnable. View return policy

Human CCL3/MIP-1 alpha DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human CCL3/MIP-1 alpha DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human CCL3/MIP-1 alpha.

Specifications

Product Type ELISA Kit
Quantity 1 Kit
Species Human
Conjugate Biotin
Assay Range 7.81 to 500 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Format 15 96-well Plates
For Use With (Application) ELISA
Gene ID (Entrez) 6348
Synonym C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.