Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCL5/RANTES Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-5316899
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2781SP 25 μg
MAB2781100 100 μg
MAB2781500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB2781SP Supplier R&D Systems Supplier No. MAB2781SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

CCL5/RANTES Monoclonal specifically detects CCL5/RANTES in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen CCL5/RANTES
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 16411
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.2-0.8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias chemokine (C-C motif) ligand 5, D17S136Enormally T-expressed, and presumably secreted, EoCP, Eosinophil chemotactic cytokine, RANTES, SISd, SIS-delta, small inducible cytokine A5 (RANTES), small inducible cytokine subfamily A (Cys-Cys), member 5, Small-inducible cytokine A5, T cell-specific protein P228, T-cell specific protein p288, TCP228T-cell-specific protein RANTES
Gene Symbols CCL5
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL5/RANTES
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6352
Test Specificity Detects human CCL5/RANTES in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human CCL1, 2, 3, 4, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, 27, 28, recombinant mouse CCL1, 2, 3, 4, 5, 6, 7, 8, 9/10/MIP-1 gamma , 11, 12, 17, 19, 20, 21, 22, 24, 25, 27, recombinant cotton rat CCL3, 4, 5, or recombinant rat CCL20.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.