Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD19 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
$151.20 - $303.50
Specifications
Antigen | CD19 |
---|---|
Clone | 4G7-2E3 |
Dilution | Flow Cytometry 5 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB4867T025
![]() |
R&D Systems
FAB4867T025 |
25 Tests |
Each for $151.20
|
|
|||||
FAB4867T100
![]() |
R&D Systems
FAB4867T100 |
100 Tests |
Each for $303.50
|
|
|||||
Description
CD19 Monoclonal specifically detects CD19 in Human samples. It is validated for Flow Cytometry.Specifications
CD19 | |
Flow Cytometry 5 uL/10^6 cells | |
Monoclonal | |
Supernatant | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
930 | |
Human CLL cells | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
4G7-2E3 | |
Flow Cytometry | |
Alexa Fluor 594 | |
Mouse | |
Human | |
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, Leu-12, MGC12802, T-cell surface antigen Leu-12 | |
CD19 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human CD19. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title