Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD3 delta/CD3d Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB10672SP
This item is not returnable.
View return policy
Description
CD3 delta Monoclonal antibody specifically detects CD3 delta in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
CD3 delta | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CD3 antigen, delta subunit, CD3D, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, T3DOKT3, delta chain, T-cell receptor T3 delta chain, T-cell surface glycoprotein CD3 delta chain | |
Human embryonic kidney cell HEK293-derived human CD3 delta/CD3d, Phe22-Ala105, Accession # P04234 | |
25 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2b |
Western Blot, Immunohistochemistry | |
1033614 | |
Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL | |
P04234 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
915 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction