Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD3 delta/CD3d Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $432.50
Specifications
Antigen | CD3 delta |
---|---|
Clone | 1033614 |
Dilution | Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10672SP
![]() |
R&D Systems
MAB10672SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10672100
![]() |
R&D Systems
MAB10672100 |
100 μg |
Each for $432.50
|
|
|||||
Description
CD3 delta Monoclonal antibody specifically detects CD3 delta in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
CD3 delta | |
Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CD3 antigen, delta subunit, CD3D, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, T3DOKT3, delta chain, T-cell receptor T3 delta chain, T-cell surface glycoprotein CD3 delta chain | |
Human embryonic kidney cell HEK293-derived human CD3 delta/CD3d, Phe22-Ala105, Accession # P04234 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1033614 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
P04234 | |
915 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title