Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD30/TNFRSF8 F(ab')2 (Research Grade Brentuximab Biosimilar) Antibody, Novus Biologicals™

Human Monoclonal Antibody
$189.00 - $464.50
Specifications
Antigen | CD30/TNFRSF8 |
---|---|
Clone | Hu8F2 |
Dilution | Flow Cytometry 0.25 ug/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB129FAB21
![]() |
R&D Systems
MAB11129FAB2100 |
100 μg |
Each for $464.50
|
|
|||||
MAB29FAB2SP
![]() |
R&D Systems
MAB11129FAB2SP |
25 μg |
Each for $189.00
|
|
|||||
Description
CD30/TNFRSF8 Monoclonal antibody specifically detects CD30/TNFRSF8 in Human samples. It is validated for Flow CytometrySpecifications
CD30/TNFRSF8 | |
Flow Cytometry 0.25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
943 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Hu8F2 | |
Flow Cytometry | |
Unconjugated | |
Human | |
Human | |
CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, TNFRSF8, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8 | |
Human CD30/TNFRSF8 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title