Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CL-P1/COLEC12 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $491.00
Specifications
Antigen | CL-P1/COLEC12 |
---|---|
Clone | 388305 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB26901SP
![]() |
R&D Systems
MAB26901SP |
25 μg |
Each for $200.50
|
|
|||||
MAB26901
![]() |
R&D Systems
MAB26901 |
100 μg |
Each for $491.00
|
|
|||||
Description
CL-P1/COLEC12 Monoclonal specifically detects CL-P1/COLEC12 in Human samples. It is validated for Western Blot.Specifications
CL-P1/COLEC12 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Human | |
Q5KU26 | |
81035 | |
Mouse myeloma cell line NS0-derived recombinant human CL-P1/COLEC12 isoform 1 Leu57-Leu742 Accession # Q5KU26 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CL-P1/COLEC12 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CL-P1 is observed. |
388305 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CL-P1, CLP1hCL-P1, COLEC12, collectin placenta 1, Collectin placenta protein 1, collectin sub-family member 12, collectin-12, NSR2, Nurse cell scavenger receptor 2, SCARA4, SCARA4NSR2, Scavenger receptor class A member 4, scavenger receptor class A, member 4, SRCL Type I, SRCLScavenger receptor with C-type lectin | |
COLEC12 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title