Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Claudin-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $479.50
Specifications
Antigen | Claudin-4 |
---|---|
Clone | 382316 |
Dilution | Immunocytochemistry 8-25 ug/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB42191SP
![]() |
R&D Systems
MAB42191SP |
25 μg |
Each for $200.50
|
|
|||||
MAB42191100
![]() |
R&D Systems
MAB42191100 |
100 μg |
Each for $479.50
|
|
|||||
Description
Claudin-4 Monoclonal specifically detects Claudin-4 in Human samples. It is validated for Immunocytochemistry.Specifications
Claudin-4 | |
Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Supernatant | |
Human | |
NP_001296 | |
1364.0 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
382316 | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
claudin 4, Claudin4, CLDN4, Clostridium perfringens enterotoxin receptor, Clostridium perfringens enterotoxin receptor 1, CPE-R, CPE-RCPETR, CPE-receptor, CPETR1, hCPE-R, WBSCR8, WBSCR8CPERCPETR1claudin-4, Williams-Beuren syndrome chromosomal region 8 protein | |
NS0 mouse myeloma cell line transfected with human Claudin-4, Met1-Val209, Accession No. NP_001296 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Claudin-4 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title