Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CLEC17A Antibody, R&D Systems™

Mouse Monoclonal Antibody
$212.50 - $485.00
Specifications
Antigen | CLEC17A |
---|---|
Clone | 1064719 |
Dilution | 0.25 μg/106 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB101471SP
![]() |
R&D Systems
MAB101471SP |
25 μg |
Each for $212.50
|
|
|||||
RD10000121
![]() |
R&D Systems
MAB101471100 |
100 μg |
Each for $485.00
|
|
|||||
Description
CLEC17A Monoclonal antibody specifically detects CLEC17A in Human samples. It is validated for Flow CytometrySpecifications
CLEC17A | |
0.25 μg/106 cells | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
C-type lectin domain family 17, member A | |
CHO-derived human CLEC17A, Lys194-Cys378, Accession # Q6ZS10 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1064719 | |
Flow Cytometry | |
Unconjugated | |
Mouse | |
Human | |
Q6ZS10 | |
388512 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title