Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Coagulation Factor III/ Tissue Factor Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
$227.50 - $584.50
Specifications
Antigen | Coagulation Factor III/Tissue Factor |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.5 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF |
Classification | Polyclonal |
Description
Coagulation Factor III/Tissue Factor Polyclonal specifically detects Coagulation Factor III/Tissue Factor in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF-ready.Specifications
Coagulation Factor III/Tissue Factor | |
Western Blot 0.5 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, CyTOF-ready | |
Polyclonal | |
Goat | |
Human | |
CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), F3, FLJ17960, TF, TFA, Thromboplastin, tissue factor | |
F3 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF | |
Unconjugated | |
RUO | |
P13726 | |
2152 | |
Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor III/Tissue Factor Gly34-Glu251 Accession # P13726 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Coagulation Factor III/Tissue Factor in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse Coagulation Factor III/Tissue Factor is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title