Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human COCO Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3047SP
This item is not returnable.
View return policy
Description
COCO/DAND5 Polyclonal specifically detects COCO/DAND5 in Human samples. It is validated for Western Blot.Specifications
COCO/DAND5 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CER2CRL2, Cerberus 2, Cerberus like-2, cerberus-like 2, Cerberus-like protein 2, cerl-2, CERL2, CKTSF1B3, CKTSF1B3MGC126849, COCO, Cysteine knot superfamily 1, BMP antagonist 3, DAN domain family member 5, DAN domain family, member 5, DAND5, Dante, DTE, FLJ38607, GREM3, GREM3cerberus 2, Gremlin-3, SP1 | |
Goat | |
Affinity Purified | |
RUO | |
199699 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
NP_689867 | |
DAND5 | |
E. coli-derived recombinant human COCO (R&D Systems, Catalog # 3047-CC) Arg23-Ala189 Accession # NP_689867 | |
25 μg | |
Primary | |
Detects human COCO in direct ELISAs and Western blots. In these formats, less than 1% cross-reactivity with recombinant mouse COCO is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction