Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human COCO Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $526.50
Specifications
Antigen | COCO/DAND5 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
COCO/DAND5 Polyclonal specifically detects COCO/DAND5 in Human samples. It is validated for Western Blot.Specifications
COCO/DAND5 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
CER2CRL2, Cerberus 2, Cerberus like-2, cerberus-like 2, Cerberus-like protein 2, cerl-2, CERL2, CKTSF1B3, CKTSF1B3MGC126849, COCO, Cysteine knot superfamily 1, BMP antagonist 3, DAN domain family member 5, DAN domain family, member 5, DAND5, Dante, DTE, FLJ38607, GREM3, GREM3cerberus 2, Gremlin-3, SP1 | |
DAND5 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
NP_689867 | |
199699 | |
E. coli-derived recombinant human COCO (R&D Systems, Catalog # 3047-CC) Arg23-Ala189 Accession # NP_689867 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human COCO in direct ELISAs and Western blots. In these formats, less than 1% cross-reactivity with recombinant mouse COCO is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title