Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Collagen I alpha 1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB62201SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB62201SP 25 μg
MAB62201100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB62201SP Supplier R&D Systems Supplier No. MAB62201SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Collagen I alpha 1 Monoclonal antibody specifically detects Collagen I alpha 1 in Human samples. It is validated for ELISA

Specifications

Antigen Collagen I alpha 1
Applications ELISA
Classification Monoclonal
Clone 816103
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Accession No. P02452
Gene Alias Alpha-1 type I collagen, COL1A1, COL-IA1, Collagen 1, Collagen 1 alpha 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, Collagen Type I Alpha 1 Chain, collagen, type I, alpha 1, Collagen1, Collagen-1, EDSARTH1, OI4, pro-alpha-1 collagen type 1, Type I Procollagen Alpha 1 Chain
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human Collagen I alpha 1, Gln23-Lys277 + Gly1094-Leu1464, Accession # P02452
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1277
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.