Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Collagen I alpha 1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $479.50
Specifications
| Antigen | Collagen I alpha 1 |
|---|---|
| Clone | 816103 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB62201SP
![]() |
R&D Systems
MAB62201SP |
25 μg |
Each for $200.50
|
|
|||||
MAB62201100
![]() |
R&D Systems
MAB62201100 |
100 μg |
Each for $479.50
|
|
|||||
Description
Collagen I alpha 1 Monoclonal antibody specifically detects Collagen I alpha 1 in Human samples. It is validated for ELISASpecifications
| Collagen I alpha 1 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Alpha-1 type I collagen, COL1A1, COL-IA1, Collagen 1, Collagen 1 alpha 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, Collagen Type I Alpha 1 Chain, collagen, type I, alpha 1, Collagen1, Collagen-1, EDSARTH1, OI4, pro-alpha-1 collagen type 1, Type I Procollagen Alpha 1 Chain | |
| Chinese Hamster Ovary cell line, CHO-derived human Collagen I alpha 1, Gln23-Lys277 + Gly1094-Leu1464, Accession # P02452 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 816103 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P02452 | |
| 1277 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title