Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Contactin-1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $467.50
Specifications
Antigen | Contactin-1 |
---|---|
Clone | 2332B |
Dilution | Western Blot 2 μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9041SP
![]() |
R&D Systems
MAB9041SP |
25 μg |
Each for $214.00
|
|
|||||
MAB9041100
![]() |
R&D Systems
MAB9041100 |
100 μg |
Each for $467.50
|
|
|||||
Description
Contactin-1 Monoclonal specifically detects Contactin-1 in Human samples. It is validated for Western Blot.Specifications
Contactin-1 | |
Western Blot 2 μg/mL | |
Monoclonal | |
Supernatant | |
Human | |
Q12860 | |
1272.0 | |
IgG | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
2332B | |
Western Blot | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CNTN1, contactin 1, contactin-1, Contactin1, F11, F3, F3cam, Glycoprotein gp135, GP135, Neural cell surface protein F3 | |
Human embryonic kidney cell, HEK293-derived human Contactin-1, Met1-Ser993, Accession No. Q12860 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Contactin-1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title