Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Contactin-4 Isoform a Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | Contactin-4 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Neutralization 5 ug/mL |
Applications | Western Blot, Neutralization |
Classification | Polyclonal |
Description
Contactin-4 Polyclonal specifically detects Contactin-4 in Human samples. It is validated for Western Blot, Neutralization.Specifications
Contactin-4 | |
Western Blot 0.1 ug/mL, Neutralization 5 ug/mL | |
Polyclonal | |
Goat | |
Human | |
AXCAM, axonal-associated cell adhesion molecule, BIG-2, BIG-2Brain-derived immunoglobulin superfamily protein 2, CNTN4, contactin 4, contactin-4, Contactin4, MGC33615, neural cell adhesion protein BIG-2 | |
CNTN4 | |
IgG | |
Antigen Affinity-purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Neutralization | |
Unconjugated | |
RUO | |
Q8IWV2 | |
152330 | |
Mouse myeloma cell line NS0-derived recombinant human Contactin-4 isoform a Asp19-Gly1001 Accession # Q8IWV2 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects Contactin-4 isoform a in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) CNTN2, recombinant mouse CNTN3, and rhCNTN5 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title