Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Coronavirus Nucleocapsid Protein Antibody (14) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB306078 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB306078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB306078 Supplier Novus Biologicals Supplier No. NBP323524
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Human Coronavirus Nucleocapsid Protein Monoclonal antibody specifically detects Human Coronavirus Nucleocapsid Protein in HCoV-NL63 samples. It is validated for ELISA

Specifications

Antigen Human Coronavirus Nucleocapsid Protein
Applications ELISA
Classification Monoclonal
Clone 14
Conjugate Unconjugated
Dilution ELISA 1:1000-1:2000
Formulation 0.2 um filtered solution in PBS
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human coronavirus (HCoV-229E) Nucleocapsid (Accession#: NP_073556.1; Met1-Asn389)
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species HCoV-NL63
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.