Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Coronavirus Nucleocapsid Protein Rabbit anti-HCoV-OC43, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB167214 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167214 20 μg
NB167213 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167214 Supplier Novus Biologicals Supplier No. NBP31605920UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Human Coronavirus Nucleocapsid Protein Polyclonal antibody specifically detects Human Coronavirus Nucleocapsid Protein in HCoV-OC43 samples. It is validated for Western Blot

Specifications

Antigen Human Coronavirus Nucleocapsid Protein
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of coronavirus Nucleoprotein (YP_009555245.1).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species HCoV-OC43
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.