Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Coronavirus Spike Protein Antibody (028)

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290334
Description
Human Coronavirus Spike Protein Monoclonal specifically detects Human Coronavirus Spike Protein in Virus samples. It is validated for ELISA.Specifications
Human Coronavirus Spike Protein | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
HKU1, Spike glycoprotein | |
This antibody was obtained from a rabbit immunized with purified, recombinant HCoV-HKU1 (isolate N1) Spike/S1 (YP_173238.1; Met1-Arg760) | |
50 μL | |
Primary | |
HCoV-HKU1 (isolate N1) Spike/S1 No cross-reactivity in ELISA with: HCoV-HKU1 (isolate N5) Spike S1+S2 ECD Protein SARS-CoV-2 Spike S1 Protein SARS-CoV-2 Spike S1 NTD Protein SARS-CoV-2 Spike S1 NTD Protein SARS-CoV-2 Spike RBD Protein SARS-CoV-2 Spike S2 Protein SARS-CoV-2 Spike S1+S2 ECD Protein SARS-CoV Spike S1+S2 ECD (S577A, Isolate Tor2) Protein MERS-CoV Spike S1+S2 ECD Protein HCoV-NL63 Spike S1+S2 ECD Protein HCoV-229E Spike S1+S2 ECD Protein HCoV-OC43 Spike S1+S2 ECD Protein | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
28 | |
ELISA 1:5000-1:10000 | |
YP_173238.1 | |
Rabbit | |
Protein A purified | |
RUO | |
3200426 | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction