Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Coronavirus Spike Protein Antibody (028)

Rabbit Monoclonal Antibody
$426.00 - $464.00
Specifications
Antigen | Human Coronavirus Spike Protein |
---|---|
Clone | 28 |
Dilution | ELISA 1:5000-1:10000 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Human Coronavirus Spike Protein Monoclonal specifically detects Human Coronavirus Spike Protein in Virus samples. It is validated for ELISA.Specifications
Human Coronavirus Spike Protein | |
ELISA 1:5000-1:10000 | |
Unconjugated | |
Rabbit | |
Virus | |
YP_173238.1 | |
3200426 | |
IgG | |
Protein A purified | |
HCoV-HKU1 (isolate N1) Spike/S1 No cross-reactivity in ELISA with: HCoV-HKU1 (isolate N5) Spike S1+S2 ECD Protein SARS-CoV-2 Spike S1 Protein SARS-CoV-2 Spike S1 NTD Protein SARS-CoV-2 Spike S1 NTD Protein SARS-CoV-2 Spike RBD Protein SARS-CoV-2 Spike S2 Protein SARS-CoV-2 Spike S1+S2 ECD Protein SARS-CoV Spike S1+S2 ECD (S577A, Isolate Tor2) Protein MERS-CoV Spike S1+S2 ECD Protein HCoV-NL63 Spike S1+S2 ECD Protein HCoV-229E Spike S1+S2 ECD Protein HCoV-OC43 Spike S1+S2 ECD Protein |
28 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
HKU1, Spike glycoprotein | |
This antibody was obtained from a rabbit immunized with purified, recombinant HCoV-HKU1 (isolate N1) Spike/S1 (YP_173238.1; Met1-Arg760) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title