Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Coronavirus Spike Protein Mouse anti-HCoV-OC43, Clone: 29, Novus Biologicals™
SDP

Catalog No. NB187991 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB187991 100 μg
NB187992 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB187991 Supplier Novus Biologicals Supplier No. NBP314658100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Human Coronavirus Spike Protein Monoclonal antibody specifically detects Human Coronavirus Spike Protein in HCoV-OC43 samples. It is validated for Western Blot, ELISA

Specifications

Antigen Human Coronavirus Spike Protein
Applications Western Blot, ELISA
Classification Monoclonal
Clone 29
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, ELISA 1:1000-1:2000
Formulation 0.2μm filtered solution in PBS without Preservative
Gene Alias HKU1, Spike glycoprotein
Host Species Mouse
Immunogen This product is a recombinant monoclonal antibody expressed from HEK293 cells.
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3200426
Target Species HCoV-OC43
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.