Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXADR Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3336
This item is not returnable.
View return policy
Description
CXADR Polyclonal specifically detects CXADR in Human samples. It is validated for Western Blot.Specifications
CXADR | |
Polyclonal | |
Unconjugated | |
P78310 | |
CXADR | |
Mouse myeloma cell line NS0-derived recombinant human CXADR Leu20-Gly237 Accession # P78310 | |
100 μg | |
Primary | |
Detects human CXADR in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse CXADR is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
CAR10Coxsackievirus B-adenovirus receptor, CAR4/6, coxsackie virus and adenovirus receptor, coxsackie virus B receptor, coxsackievirus and adenovirus receptor, CVB3 binding protein, CVB3 BP, CVB3-binding protein, hCAR, HCVADR | |
Goat | |
Affinity Purified | |
RUO | |
1525 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction