Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXADR Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $514.50
Specifications
Antigen | CXADR |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
CXADR Polyclonal specifically detects CXADR in Human samples. It is validated for Western Blot.Specifications
CXADR | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
CAR10Coxsackievirus B-adenovirus receptor, CAR4/6, coxsackie virus and adenovirus receptor, coxsackie virus B receptor, coxsackievirus and adenovirus receptor, CVB3 binding protein, CVB3 BP, CVB3-binding protein, hCAR, HCVADR | |
CXADR | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P78310 | |
1525 | |
Mouse myeloma cell line NS0-derived recombinant human CXADR Leu20-Gly237 Accession # P78310 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human CXADR in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse CXADR is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title