Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCL9/MIG Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB392500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB392500 500 μg
MAB392SP 25 μg
MAB392100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB392500 Supplier R&D Systems Supplier No. MAB392500
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 14 publications

CXCL9/MIG Monoclonal specifically detects CXCL9/MIG in Human samples. It is validated for Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CXCL9/MIG
Applications Western Blot, Neutralization, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 49106
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 4-20 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q07325
Gene Alias chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIG, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma
Gene Symbols CXCL9
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL9/MIG Thr23-Thr125 Accession # Q07325
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4283
Test Specificity Detects human CXCL9/MIG in ELISAs and Western blots. In ELISAs, does not cross-react with recombinant mouse (rm) CXCL9, recombinant human CXCL10.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.