Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCL9/MIG Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 14 publications
$200.16 - $2171.79
Specifications
| Antigen | CXCL9/MIG |
|---|---|
| Clone | 49106 |
| Dilution | Western Blot 1 ug/mL, Neutralization 4-20 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Neutralization, Flow Cytometry, Immunocytochemistry, CyTOF |
| Classification | Monoclonal |
Description
CXCL9/MIG Monoclonal specifically detects CXCL9/MIG in Human samples. It is validated for Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
| CXCL9/MIG | |
| Western Blot 1 ug/mL, Neutralization 4-20 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q07325 | |
| 4283 | |
| E. coli-derived recombinant human CXCL9/MIG Thr23-Thr125 Accession # Q07325 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CXCL9/MIG in ELISAs and Western blots. In ELISAs, does not cross-react with recombinant mouse (rm) CXCL9, recombinant human CXCL10. |
| 49106 | |
| Western Blot, Neutralization, Flow Cytometry, Immunocytochemistry, CyTOF | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIG, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma | |
| CXCL9 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title