Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCR7/RDC-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $549.50
Specifications
Antigen | CXCR7/RDC-1 |
---|---|
Clone | 11G8R |
Dilution | Flow Cytometry 0.25 μg/mL, Immunohistochemistry 5-25 μg/mL, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB42273RSP
![]() |
R&D Systems
MAB42273RSP |
25 μg |
Each for $214.00
|
|
|||||
MA42273R100
![]() |
R&D Systems
MAB42273R100 |
100 μg |
Each for $549.50
|
|
|||||
Description
CXCR7/RDC-1 Monoclonal specifically detects CXCR7/RDC-1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
CXCR7/RDC-1 | |
Flow Cytometry 0.25 μg/mL, Immunohistochemistry 5-25 μg/mL, CyTOF-ready | |
Monoclonal | |
Supernatant | |
Human | |
P25106 | |
57007 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
11G8R | |
Flow Cytometry, Immunohistochemistry, CyTOF | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
ACKR3, chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CMKOR1, CXCR7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog | |
Human CXCR7 encoding plasmid Accession # P25106 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CXCR7/RDC-1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title