Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Cystatin A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7064367
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1407SP 25 μg
MAB1407 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1407SP Supplier R&D Systems Supplier No. MAB1407SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cystatin A Monoclonal specifically detects Cystatin A in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Cystatin A
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 224705
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P01040
Gene Alias AREI, CSTA, cystatin A (stefin A), cystatin AS, cystatin-A, Stefin A, STF1, STF1Cystatin-AS, STFA, STFAStefin-A
Gene Symbols CSTA
Host Species Mouse
Immunogen E. coli-derived recombinant human Cystatin A Ile2-Phe98 Accession # P01040
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1475
Test Specificity Detects human Cystatin A in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) Cystatins B, C, D, E/M, F, S, SA, rhFetuin A, rhFetuin B, rhHPRG, rhKininogen, or rhKininostatin is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.