Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Dectin-2/CLEC6A Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $479.50
Specifications
| Antigen | Dectin-2/CLEC6A |
|---|---|
| Clone | 545925 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB31141SP
![]() |
R&D Systems
MAB31141SP |
25 μg |
Each for $200.50
|
|
|||||
MAB31141100
![]() |
R&D Systems
MAB31141100 |
100 μg |
Each for $479.50
|
|
|||||
Description
Dectin-2/CLEC6A Monoclonal specifically detects Dectin-2/CLEC6A in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| Dectin-2/CLEC6A | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CLEC4N, CLEC6A, CLECSF10, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 10, C-type lectin domain family 6 member A, C-type lectin domain family 6, member A, C-type lectin superfamily member 10, DC-associated C-type lectin 2, dectin 2, Dectin-2, DECTIN2, Dendritic cell-associated C-type lectin 2 | |
| Mouse myeloma cell line NS0-derived recombinant human Dectin-2/CLEC6A, Thr46-Leu209, Accession # Q6EIG7 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 545925 | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q6EIG7 | |
| 93978 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title