Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Desmin Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems MAB11607100
This item is not returnable.
View return policy
Description
Desmin Monoclonal antibody specifically detects Desmin in Human samples. It is validated for Immunocytochemistry,Western Blot,Simple Western,Immunohistochemistry,Multiplex ImmunofluorescenceSpecifications
Desmin | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
CMD1I, CMD1IFLJ41013, CSM1, CSM2, DES, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein | |
E. coli derived recombinant human Desmin, Accession # P17661 | |
100 μg | |
Primary | |
Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. | |
IgG2a |
Immunocytochemistry, Western Blot, Simple Western, Immunohistochemistry, Multiplex | |
1089115 | |
Western Blot 1 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL, Multiplex Immunofluorescence 1 μg/mL | |
P17661 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1674 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction