Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Desmin Antibody, R&D Systems™
Mouse Monoclonal Antibody
$205.85 - $485.42
Specifications
Antigen | Desmin |
---|---|
Clone | 1089115 |
Dilution | Western Blot 1 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL, Multiplex Immunofluorescence 1 μg/mL |
Applications | Immunocytochemistry, Western Blot, Simple Western, Immunohistochemistry, Multiplex |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11607SP
|
R&D Systems
MAB11607SP |
25 μg |
Each for $205.85
|
|
|||||
MAB11607100
|
R&D Systems
MAB11607100 |
100 μg |
Each for $485.42
|
|
|||||
Description
Desmin Monoclonal antibody specifically detects Desmin in Human samples. It is validated for Immunocytochemistry,Western Blot,Simple Western,Immunohistochemistry,Multiplex ImmunofluorescenceSpecifications
Desmin | |
Western Blot 1 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL, Multiplex Immunofluorescence 1 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
CMD1I, CMD1IFLJ41013, CSM1, CSM2, DES, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein | |
E. coli derived recombinant human Desmin, Accession # P17661 | |
Primary | |
Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
1089115 | |
Immunocytochemistry, Western Blot, Simple Western, Immunohistochemistry, Multiplex | |
Unconjugated | |
Mouse | |
Human | |
P17661 | |
1674 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title