Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Desmoglein-3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1720
This item is not returnable.
View return policy
Description
Desmoglein-3 Polyclonal specifically detects Desmoglein-3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Desmoglein-3 | |
Polyclonal | |
Unconjugated | |
P32926 | |
DSG3 | |
Mouse myeloma cell line NS0-derived recombinant human Desmoglein-3 Glu50-Arg615 Accession # P32926 | |
100 μg | |
Primary | |
Detects human Desmoglein-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) Desmoglein-1 and rhDesmoglein-2 is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Cadherin family member 6, CDHF6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, Desmoglein3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, DSG3, pemphigus vulgaris antigen, PVA | |
Goat | |
Affinity Purified | |
RUO | |
1830 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction