Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Desmoglein-3 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $468.50
Specifications
Antigen | Desmoglein-3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Description
Desmoglein-3 Polyclonal specifically detects Desmoglein-3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Desmoglein-3 | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Human | |
Cadherin family member 6, CDHF6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, Desmoglein3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, DSG3, pemphigus vulgaris antigen, PVA | |
DSG3 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
P32926 | |
1830 | |
Mouse myeloma cell line NS0-derived recombinant human Desmoglein-3 Glu50-Arg615 Accession # P32926 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Desmoglein-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) Desmoglein-1 and rhDesmoglein-2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title