Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Dopamine D2 R/DRD2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $549.50
Specifications
Antigen | Dopamine D2R/DRD2 |
---|---|
Clone | 960710 |
Dilution | Immunohistochemistry 5-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9266SP
![]() |
R&D Systems
MAB9266SP |
25 μg |
Each for $214.00
|
|
|||||
MAB9266100
![]() |
R&D Systems
MAB9266100 |
100 μg |
Each for $549.50
|
|
|||||
Description
Dopamine D2R/DRD2 Monoclonal specifically detects Dopamine D2R/DRD2 in Human samples. It is validated for Immunohistochemistry.Specifications
Dopamine D2R/DRD2 | |
Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P14416 | |
1813 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
960710 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
D(2) dopamine receptor, D2DR, D2R, Dopamine D2 receptor, Dopamine D2R, dopamine receptor D2, dopamine receptor D2 isoform, DRD2, seven transmembrane helix receptor | |
Human DRD2 synthetic peptide Accession # P14416 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Dopamine D2 R/DRD2 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title