Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Dopamine D5R/DRD5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB8286
This item is not returnable.
View return policy
Description
Dopamine D5R/DRD5 Monoclonal specifically detects Dopamine D5R/DRD5 in Human samples. It is validated for Immunohistochemistry.Specifications
Dopamine D5R/DRD5 | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
P21918 | |
DRD5 | |
NS0 mouse myeloma cell line transfected with human Dopamine D5 R/DRD5 Accession # P21918 | |
100 μg | |
Primary | |
Detects HEK293 cells transfected with human Dopamine D5 R/DRD5 by Flow Cytometry. Does not detect untransfected or irrelevant transfected HEK293 cells. | |
Human | |
Purified |
Immunohistochemistry | |
889040 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
D(1B) dopamine receptor, D(5) dopamine receptor, D1beta dopamine receptor, DBDR, Dopamine D5 R, Dopamine D5 receptor, dopamine receptor D1B, dopamine receptor D5, DRD1B, DRD1BDBDR, DRD1L2, DRD1L2MGC10601, DRD5 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1816 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction