Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Dopamine D5R/DRD5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | Dopamine D5R/DRD5 |
---|---|
Clone | 889040 |
Concentration | LYOPH |
Dilution | Immunohistochemistry 8-25 ug/mL |
Applications | Immunohistochemistry |
Description
Dopamine D5R/DRD5 Monoclonal specifically detects Dopamine D5R/DRD5 in Human samples. It is validated for Immunohistochemistry.Specifications
Dopamine D5R/DRD5 | |
LYOPH | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
P21918 | |
1816 | |
NS0 mouse myeloma cell line transfected with human Dopamine D5 R/DRD5 Accession # P21918 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects HEK293 cells transfected with human Dopamine D5 R/DRD5 by Flow Cytometry. Does not detect untransfected or irrelevant transfected HEK293 cells. |
889040 | |
Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
D(1B) dopamine receptor, D(5) dopamine receptor, D1beta dopamine receptor, DBDR, Dopamine D5 R, Dopamine D5 receptor, dopamine receptor D1B, dopamine receptor D5, DRD1B, DRD1BDBDR, DRD1L2, DRD1L2MGC10601, DRD5 | |
DRD5 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title