Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human E-Selectin/CD62E Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $561.50
Specifications
Antigen | E-Selectin/CD62E |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Adhesion Blockade |
Applications | Western Blot, Adhesion Blockade |
Classification | Polyclonal |
Description
E-Selectin/CD62E Polyclonal specifically detects E-Selectin/CD62E in Human samples. It is validated for Western Blot, Adhesion Blockade.Specifications
E-Selectin/CD62E | |
Western Blot 0.1 ug/mL, Adhesion Blockade | |
Polyclonal | |
Sheep | |
Human | |
6401 | |
Chinese hamster ovary cell line CHO-derived recombinant human E-Selectin | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Selected for its ability to inhibit the block U937 cell adhesion to immobilized rhE-Selectin. In direct ELISAs, less than 5% cross-reactivity with rmE-Selectin and rrE-Selectin is observed and less than 1% cross-reactivity with rhP-Selectin and rhL-Selectin is observed. |
LYOPH | |
Western Blot, Adhesion Blockade | |
Unconjugated | |
RUO | |
CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM1, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, SELE, selectin E | |
SELE | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title