Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EAAT1/GLAST-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | EAAT1/GLAST-1/SLC1A3 |
---|---|
Clone | 482420 |
Dilution | Immunohistochemistry 5-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB6048SP
![]() |
R&D Systems
MAB6048SP |
25 μg |
Each for $200.50
|
|
|||||
MAB6048100
![]() |
R&D Systems
MAB6048100 |
100 μg |
Each for $467.50
|
|
|||||
Description
EAAT1/GLAST-1/SLC1A3 Monoclonal specifically detects EAAT1/GLAST-1/SLC1A3 in Human samples. It is validated for Immunohistochemistry.Specifications
EAAT1/GLAST-1/SLC1A3 | |
Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
EA6, EA6FLJ25094, EAAT1GLAST-1, excitatory amino acid transporter 1, GLAST1, GLAST-1, GLASTGLAST1, SLC1A3, Sodium-dependent glutamate/aspartate transporter 1, solute carrier family 1 (glial high affinity glutamate transporter), member 3, Solute carrier family 1 member 3 | |
E. coli-derived recombinant human EAAT1/GLAST-1, His146-Ser237, Accession # P43003 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human EAAT1/GLAST-1 in direct ELISAs. |
482420 | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
P43003 | |
6507 | |
IgG2b | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title