Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ECE-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB17842
This item is not returnable.
View return policy
Description
ECE-1 Monoclonal specifically detects ECE-1 in Human samples. It is validated for Immunohistochemistry.Specifications
ECE-1 | |
Monoclonal | |
LYOPH | |
Immunohistochemistry | |
P42892 | |
ECE1 | |
Mouse myeloma cell line NS0-derived recombinant human ECE-1 Gln90-Trp770 Accession # P42892 | |
100 μg | |
Primary | |
Detects human ECE-1 in direct ELISAs. | |
Human | |
Purified |
Immunohistochemistry | |
303906 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.24, EC 3.4.24.71, ECE, ECE1, ECE-1, endothelin converting enzyme 1, endothelin-converting enzyme 1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1889 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction