Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ECE-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | ECE-1 |
---|---|
Clone | 303906 |
Concentration | LYOPH |
Dilution | Immunohistochemistry |
Applications | Immunohistochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB17842SP
![]() |
R&D Systems
MAB17842SP |
25 μg |
Each for $200.50
|
|
|||||
MAB17842
![]() |
R&D Systems
MAB17842 |
100 μg |
Each for $468.00
|
|
|||||
Description
ECE-1 Monoclonal specifically detects ECE-1 in Human samples. It is validated for Immunohistochemistry.Specifications
ECE-1 | |
LYOPH | |
Immunohistochemistry | |
Unconjugated | |
Rat | |
Human | |
P42892 | |
1889 | |
Mouse myeloma cell line NS0-derived recombinant human ECE-1 Gln90-Trp770 Accession # P42892 | |
Primary | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Detects human ECE-1 in direct ELISAs. |
303906 | |
Immunohistochemistry | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.24, EC 3.4.24.71, ECE, ECE1, ECE-1, endothelin converting enzyme 1, endothelin-converting enzyme 1 | |
ECE1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title