Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EDA/Ectodysplasin Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $667.00
Specifications
| Antigen | EDA/Ectodysplasin |
|---|---|
| Clone | 174806 |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Classification | Monoclonal |
Description
EDA/Ectodysplasin Monoclonal specifically detects EDA/Ectodysplasin in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| EDA/Ectodysplasin | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ectodysplasin A, ED1, EDAA1, EDAA2, Tabby, XHED, XLHED | |
| EDA | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 174806 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_001005609 | |
| 1896 | |
| Mouse myeloma cell line NS0-derived recombinant human EDA-A2/Ectodysplasin A2 Ala179-Ser389 Accession # NP_001005609 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human EDA/Ectodysplasin (EDA-A1 and EDA-A2) in ELISAs. In sandwich immunoassays, approximately 50% cross-reactivity with recombinant mouse EDA is observed, and no cross-reactivity with recombinant human (rh) TNF-alpha , rhXEDAR, or rhEDAR is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title