Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EGR1 Antibody, R&D Systems™

Rat Monoclonal Antibody
$208.00 - $509.00
Specifications
Antigen | EGR1 |
---|---|
Clone | 406910 |
Dilution | Immunohistochemistry 5-25 μ/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB28181SP
![]() |
R&D Systems
MAB28181SP |
25 μg |
Each for $208.00
|
|
|||||
MAB28181100
![]() |
R&D Systems
MAB28181100 |
100 μg |
Each for $509.00
|
|
|||||
Description
EGR1 Monoclonal specifically detects EGR1 in Human samples. It is validated for Immunohistochemistry.Specifications
EGR1 | |
Immunohistochemistry 5-25 μ/mL | |
Monoclonal | |
Rat | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
AT225Transcription factor ETR103, early growth response 1, early growth response protein 1, EGR-1, G0S30, KROX24, KROX-24, Nerve growth factor-induced protein A, NGFIA, NGFI-ATranscription factor Zif268, TIS8, ZIF-268, Zinc finger protein Krox-24, ZNF225, ZNF225Zinc finger protein 225 | |
E. coli-derived recombinant human EGR1, Gln282-Ser433, Accession No. P18146 | |
Primary | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
406910 | |
Immunohistochemistry | |
Unconjugated | |
Human | |
P18146 | |
1958 | |
IgG2a | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human EGR1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title