Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EphA1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $643.50
Specifications
Antigen | EphA1 |
---|---|
Clone | 86810 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
EphA1 Monoclonal specifically detects EphA1 in Human samples. It is validated for Western Blot.Specifications
EphA1 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.10, EC 2.7.10.1, EPH receptor A1, eph tyrosine kinase 1, EphA1, EPHephrin type-A receptor 1, EPHT, EPHT1erythropoietin-producing hepatoma amplified sequence, Esk, MGC163163, oncogene EPH, soluble EPHA1 variant 1, soluble EPHA1 variant 2, Tyrosine-protein kinase receptor EPH | |
EPHA1 | |
IgG2a | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
86810 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P21709 | |
2041 | |
Mouse myeloma cell line NS0-derived recombinant human EphA1 Arg24-Glu547 (predicted) Accession # P21709 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human EphA1 in direct ELISAs and Western blots. No cross-reactivity with recombinant mouse EphA2, A3, A4, A6, A7, A8, B2, B4, B5, B6, recombinant rat (rr) EphA5, rrEphB1, or recombinant human EphB1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title