Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EphA10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5188
This item is not returnable.
View return policy
Description
EphA10 Monoclonal specifically detects EphA10 in Human samples. It is validated for Immunohistochemistry.Specifications
EphA10 | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
Q5JZY3 | |
EPHA10 | |
E. coli-derived recombinant human EphA10 Glu34-Glu283 (predicted) Accession # Q5JZY3 | |
100 μg | |
Primary | |
Detects human EphA10 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human EphA1, A2, A5, A6, B2, B3, B4, B6, recombinant mouse EphA3, A4, or A8 is observed. | |
Human | |
Purified |
Immunohistochemistry | |
494130 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.10.1, EPH receptor A10, EphA10s protein, ephrin type-A receptor 10, FLJ16103, FLJ33655, MGC43817 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
284656 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction