Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EphA10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | EphA10 |
---|---|
Clone | 494130 |
Concentration | LYOPH |
Dilution | Immunohistochemistry 8-25 ug/mL |
Applications | Immunohistochemistry |
Description
EphA10 Monoclonal specifically detects EphA10 in Human samples. It is validated for Immunohistochemistry.Specifications
EphA10 | |
LYOPH | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
Q5JZY3 | |
284656 | |
E. coli-derived recombinant human EphA10 Glu34-Glu283 (predicted) Accession # Q5JZY3 | |
Primary | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Detects human EphA10 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human EphA1, A2, A5, A6, B2, B3, B4, B6, recombinant mouse EphA3, A4, or A8 is observed. |
494130 | |
Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.10.1, EPH receptor A10, EphA10s protein, ephrin type-A receptor 10, FLJ16103, FLJ33655, MGC43817 | |
EPHA10 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title