Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ErbB4/Her4 Antibody, R&D Systems™

Goat Antibody
Supplier: R&D Systems AF10446SP
This item is not returnable.
View return policy
Description
ErbB4/Her4 specifically detects ErbB4/Her4 in Human samples. It is validated for Western Blot, Knockout Validated.Specifications
ErbB4/Her4 | |
Polyclonal | |
Western Blot 1 ug/mL, Knockout Validated | |
Q15303 | |
Goat | |
Antigen Affinity-purified | |
RUO | |
2066 | |
Human | |
Purified |
Western Blot, KnockDown | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4, EC 2.7.10, EC 2.7.10.1, HER4, HER4MGC138404, p180erbB4, Proto-oncogene-like protein c-ErbB-4, receptor tyrosine-protein kinase erbB-4, Tyrosine kinase-type cell surface receptor HER4, v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 4, v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian) | |
E. coli-derived recombinant human ErbB4/Her4, Ala1027-Pro1292, Accession # Q15303 | |
25 μg | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction