Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ErbB4/Her4 Antibody, R&D Systems™

Goat Antibody
$200.50 - $468.50
Specifications
Antigen | ErbB4/Her4 |
---|---|
Dilution | Western Blot 1 ug/mL, Knockout Validated |
Applications | Western Blot, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AF10446SP
![]() |
R&D Systems
AF10446SP |
25 μg |
Each for $200.50
|
|
|||||
AF10446100
![]() |
R&D Systems
AF10446100 |
100 μg |
Each for $468.50
|
|
|||||
Description
ErbB4/Her4 specifically detects ErbB4/Her4 in Human samples. It is validated for Western Blot, Knockout Validated.Specifications
ErbB4/Her4 | |
Western Blot, KnockDown | |
Unconjugated | |
Goat | |
Human | |
Q15303 | |
2066 | |
IgG | |
Antigen Affinity-purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Western Blot 1 ug/mL, Knockout Validated | |
Polyclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4, EC 2.7.10, EC 2.7.10.1, HER4, HER4MGC138404, p180erbB4, Proto-oncogene-like protein c-ErbB-4, receptor tyrosine-protein kinase erbB-4, Tyrosine kinase-type cell surface receptor HER4, v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 4, v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian) | |
E. coli-derived recombinant human ErbB4/Her4, Ala1027-Pro1292, Accession # Q15303 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title